4-Amino-5-chloro-2,6-dimethylpyrimidine ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A151267-1G A151267-25G A151267-5G
ALA151267-1GEA 15.11 USD
ALA151267-1G ALA151267-25G ALA151267-5G
4-Amino-5-chloro-2,6-dimethylpyrimidine ≥96%
4-Amino-5-chloro-2,6-dimethylpyrimidine

4-Amino-5-chloro-2,6-dimethylpyrimidine (ACDP) is an aminopyrimidine derivative. Its interaction as an electron donor with a σ-electron acceptor, iodine has been studied. 4-Amino-5-chloro-2,6-dimethylpyrimidine participates in the synthesis of diazino[c]quinolin-5(6H)-one and -isoquinolin-6(5H)-one, diazino[c]naphthyridin-6(5H)- and -5(6H)-one derivatives. 4-Amino-5-chloro-2,6-dimethylpyrimidine is suitable reagent employed as a nucleobase standard in the systematic quantitative structure-retention relationship (QSRR) study on pyrimidines between the molecular descriptors (variables) and their chromatographic retention (experimental unit).

Formula: C₆H₈ClN₃
Melting Pt: 164-166°C
MDL Number: MFCD00191727
CAS Number: 2858-20-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7e15473862b23c0f1eab60757ac7946f2009d4e&vid=c7e15473862b23c0f1eab60757ac7946f2009d4e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR