3-Amino-5-methylthio-1H-1,2,4-triazole ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151264-25G A151264-5G
ALA151264-25GEA 74.01 USD
ALA151264-25G ALA151264-5G
3-Amino-5-methylthio-1H-1,2,4-triazole ≥97% (by HPLC)
3-Amino-5-methylthio-1H-1,2,4-triazole

3-Amino-5-methylthio-1H-1,2,4-triazole was used in the synthesis of heterocyclic disperse azo dyes based on 8-hydroxyquinoline.

Formula: C₃H₆N₄S
Melting Pt: 130-139°C
MDL Number: MFCD00005232
CAS Number: 45534-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf4ca2cc4499257e4c2cb7502218876a8b563899&vid=bf4ca2cc4499257e4c2cb7502218876a8b563899&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR