4-Amino-2,5-xylenol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-2,5-dimethylphenol

A151261-100G A151261-1G A151261-25G A151261-5G
ALA151261-100GEA 1426.81 USD
ALA151261-100G ALA151261-1G ALA151261-25G ALA151261-5G
4-Amino-2,5-xylenol ≥97%
4-Amino-2,5-xylenol

4-Amino-2,5-dimethylphenol may be used in chemical synthesis studies.

Formula: C₈H₁₁NO
MW: 137.1800 g/mol
Melting Pt: 242-243°C
Storage Temperature: Ambient
MDL Number: MFCD00007873
CAS Number: 3096-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e2ac3c09149b385c17fae00518d0a7cf006829&vid=a0e2ac3c09149b385c17fae00518d0a7cf006829&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR