4-Aminoquinoline ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151254-250MG A151254-5G
ALA151254250MGEA 26.51 USD
ALA151254250MG ALA151254-5G
4-Aminoquinoline ≥98%
4-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00463448
CAS Number: 578-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd4b14c7fd586619e25e4dfa962802e88a1d63e4&vid=fd4b14c7fd586619e25e4dfa962802e88a1d63e4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR