2-Amino-5-nitrophenol ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Hydroxy-4-nitroaniline

A151253-250G A151253-25G A151253-5G
ALA151253-250GEA 129.11 USD
ALA151253-250G ALA151253-25G ALA151253-5G
2-Amino-5-nitrophenol ≥98% (by HPLC)
2-Amino-5-nitrophenol

2-Amino-5-nitrophenol was used: . as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides . as semipermanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products ? in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents Reactant for: . Diazotation and coupling reactions . Preparation of biologically and pharmacologically active molecules

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 198-202 °C
MDL Number: MFCD00007692
CAS Number: 121-88-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1360ab296d5a572f56ebb335f9bec6cb111c49e&vid=e1360ab296d5a572f56ebb335f9bec6cb111c49e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR