1-Acetyl-1,2,3,4-tetrahydroquinoline 98 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1,2,3,4-Tetrahydro-1-acetylquinoline

A151251-100G A151251-1G A151251-25G A151251-5G
ALA151251-100GEA 532.86 USD
ALA151251-100G ALA151251-1G ALA151251-25G ALA151251-5G
1-Acetyl-1,2,3,4-tetrahydroquinoline 98 GC_ASSAY_METHOD
1-Acetyl-1,2,3,4-tetrahydroquinoline
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 295°C
MDL Number: MFCD01195305
CAS Number: 4169-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc7370aec1d2e0337f2d046063e4cef8f842aa53&vid=cc7370aec1d2e0337f2d046063e4cef8f842aa53&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR