6-Amino-2-methylquinoline ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-methylquinolin-6-amine

A151250-1G A151250-250MG
ALA151250-1GEA 109.16 USD
ALA151250-1G ALA151250250MG
6-Amino-2-methylquinoline ≥98%
6-Amino-2-methylquinoline

6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 189-191°C
Storage Temperature: Ambient
MDL Number: MFCD00052600
CAS Number: 65079-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c83d3bb2edee38bea1f946a9d813223d38bd800f&vid=c83d3bb2edee38bea1f946a9d813223d38bd800f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR