3-Aminoquinoline ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Quinolin-3-amine, Quinolin-3-ylamine

A151247-25G A151247-5G A151247-100G A151247-1G
ALA151247-25GEA 173.76 USD
ALA151247-25G ALA151247-5G ALA151247-100G ALA151247-1G
3-Aminoquinoline ≥97% (by GC)
3-Aminoquinoline

3-Aminoquinoline was used: . in liquid matrix for matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) for glycopeptides, carbohydrates and phosphopeptides . in liquid matrix during novel glycan-labeling method for quantitative profiling of N-glycan by MALDI-MS analysis . as derivatizing reagent for MALDI-MS analysis of oligosachharides

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 137 °C
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00006772
CAS Number: 580-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b339f203b49f0f9ab027fe7079602dbc25d3a7e&vid=9b339f203b49f0f9ab027fe7079602dbc25d3a7e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR