2-Amino-4-chloro-6-nitrophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151244-100G A151244-1G A151244-250MG A151244-25G A151244-5G
ALA151244-100GEA 90.16 USD
ALA151244-100G ALA151244-1G ALA151244250MG ALA151244-25G ALA151244-5G
2-Amino-4-chloro-6-nitrophenol ≥98%
2-Amino-4-chloro-6-nitrophenol
Formula: C₆H₅ClN₂O₃
MW: 188.57 g/mol
Melting Pt: 152°C
MDL Number: MFCD00035925
CAS Number: 6358-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR