4-Amino-2-chlorophenol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151243-100G A151243-1G A151243-25G A151243-5G
ALA151243-100GEA 227.91 USD
ALA151243-100G ALA151243-1G ALA151243-25G ALA151243-5G
4-Amino-2-chlorophenol ≥98% (by GC)
4-Amino-2-chlorophenol

4-Amino-2-chlorophenol (4A2CP) is a chlorinated aniline. Nephrotoxic potential of 4-amino-2-chlorophenol and its comparison with 4-aminophenol, 4-amino-2-chlorophenol, 4-amino-3-chlorophenol and 4-amino-2,6-dichlorophenol using isolated renal cortical cells from male Fischer 344 rats has been investigated.-Amino-2-chlorophenol may be used in the synthesis of the following compounds: . N-(3-chloro-4-hydroxyphenyl)-N′,N′-dimethylurea . 1,4-diketo-3-((4-[N-(3-chloro-4-hydroxyphenyl)amino]sulfonyl)phenyl)-6-phenylpyrrolo[3,4-c]pyrrole, a novel fluorescent pH-i

Formula: C₆H₆ClNO
Melting Pt: 150-153°C
Storage Temperature: Refrigerator
MDL Number: MFCD00128891
CAS Number: 3964-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=666b84a9cd9eb4dc7280b7ce8ea7bc11851274ab&vid=666b84a9cd9eb4dc7280b7ce8ea7bc11851274ab&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR