4-Amino-2-nitrophenol ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151242-100G
A151242-25G
A151242-5G
ALA151242-100GEA
205.11
USD
ALA151242-100G
ALA151242-25G
ALA151242-5G
4-Amino-2-nitrophenol ≥95%
4-Amino-2-nitrophenol
4-Amino-2-nitrophenol is the major metabolite of 2,4-dinitrophenol[1]. It is commonly used as semipermanent (nonoxidative) hair colorant and toner in permanent (oxidative) hair dye products.
Formula:
C₆H₆N₂O₃ Melting Pt: 125-127°C |
MDL Number:
MFCD00007876 CAS Number: 119-34-6 UN: 2811 ADR: 6.1,I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...