2-Amino-4-chloro-5-nitrophenol ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

A151241-250G A151241-25G A151241-100G
ALA151241-250GEA 496.76 USD
ALA151241-250G ALA151241-25G ALA151241-100G
2-Amino-4-chloro-5-nitrophenol ≥98% (by titration)
2-Amino-4-chloro-5-nitrophenol
Formula: C₆H₅ClN₂O₃
Melting Pt: 220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010300
CAS Number: 6358-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR