1-Aminopyrrole 98 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1H-Pyrrol-1-amine

A151234-1G A151234-250MG A151234-5G
ALA151234-1GEA 42.66 USD
ALA151234-1G ALA151234250MG ALA151234-5G
1-Aminopyrrole 98 GC_ASSAY_METHOD
1-Aminopyrrole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 174°C
Flash Pt: 64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059931
CAS Number: 765-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e36258aee4087b5721f17b6ffc73dc1ac464e9fa&vid=e36258aee4087b5721f17b6ffc73dc1ac464e9fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR