Allylurea ≥98% (by total nitrogen basis)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N-Allylurea

A151231-25G A151231-5G
ALA151231-25GEA 60.71 USD
ALA151231-25G ALA151231-5G
Allylurea ≥98% (by total nitrogen basis)
Allylurea
Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 83-88°C
Storage Temperature: Refrigerator
MDL Number: MFCD00007954
CAS Number: 557-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1833f8050e5778d83eb9014b6f385e4570c57a99&vid=1833f8050e5778d83eb9014b6f385e4570c57a99&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR