1-Acetyl-3-methylurea ≥96%

Supplier: ALADDIN SCIENTIFIC

A151230-100G A151230-25G A151230-5G
ALA151230-100GEA 1131.36 USD
ALA151230-100G ALA151230-25G ALA151230-5G
1-Acetyl-3-methylurea ≥96%
1-Acetyl-3-methylurea
Formula: C₄H₈N₂O₂
Melting Pt: 180 °C
MDL Number: MFCD00026167
CAS Number: 623-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=889a373f98ebf8f81df989591d971cdb92ea3f42&vid=889a373f98ebf8f81df989591d971cdb92ea3f42&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR