2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane ≥98% stabilized

Supplier: ALADDIN SCIENTIFIC
Warning


Stabilization: Stabilised with phenothiazine

A151228-1G A151228-250MG A151228-5G
ALA151228-1GEA 121.51 USD
ALA151228-1G ALA151228250MG ALA151228-5G
2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane ≥98% stabilized
2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane
Formula: C₈H₁₅BO₂
Density: 0.93 g/cm³
Flash Pt: 56 °C
Storage Temperature: Refrigerator
CAS Number: 911482-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee86d1dac015823d604ab03565344c13e54327f7&vid=ee86d1dac015823d604ab03565344c13e54327f7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR