ɑ-Methyl-γ-butyrolactone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Methyldihydrofuran-2(3H)-one, Dihydro-3-methyl-2(3H)-furanone, α-Methyl-γ-butyrolactone

A151225-1G A151225-5G
ALA151225-1GEA 19.86 USD
ALA151225-1G ALA151225-5G
ɑ-Methyl-γ-butyrolactone ≥98% (by GC)
alpha-Methyl-gamma-butyrolactone

α-Methyl-γ-butyrolactone undergoes benzylation to give racemic α-benzyl-α-methyl-γ-butyrolactone.α-Methyl-γ-butyrolactone was used as model compound in Bracketing experiments to investigate the thermodynamically favored site of reaction of pilocarpine

Formula: C₅H₈O₂
Boiling Pt: 78-81 °C
Density: 1.06 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005396
CAS Number: 1679-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e4146475a389d161c875b72f7e37be03ab14f4&vid=c6e4146475a389d161c875b72f7e37be03ab14f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR