4-(3,4-Dichlorophenyl)thiazol-2-amine ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

A151221-1G A151221-250MG A151221-5G
ALA151221-1GEA 45.51 USD
ALA151221-1G ALA151221250MG ALA151221-5G
4-(3,4-Dichlorophenyl)thiazol-2-amine ≥98% (by GC, titration analysis)
4-(3,4-Dichlorophenyl)thiazol-2-amine
Formula: C₉H₆Cl₂N₂S
Melting Pt: 163 °C
MDL Number: MFCD00466323
CAS Number: 39893-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a01ab45e1cf40318d0b67ae5f682d292159db80&vid=6a01ab45e1cf40318d0b67ae5f682d292159db80&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR