2-Amino-4-(1-naphthyl)thiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151220-1G A151220-250MG
ALA151220-1GEA 44.56 USD
ALA151220-1G ALA151220250MG
2-Amino-4-(1-naphthyl)thiazole ≥97%
2-Amino-4-(1-naphthyl)thiazole
Formula: C₁₃H₁₀N₂S
Melting Pt: 158 °C
MDL Number: MFCD00236016
CAS Number: 56503-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec4cdbf90deb44c08c22ca39e90f1e507b2e3687&vid=ec4cdbf90deb44c08c22ca39e90f1e507b2e3687&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR