4-(1,3-Benzothiazol-2-yl)phenylamine ≥97% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(2-Benzothiazolyl)aniline, 4-(Benzo[d]thiazol-2-yl)aniline, 2-(4-Aminophenyl)benzothiazole

A151218-1G A151218-5G
ALA151218-1GEA 19.86 USD
ALA151218-1G ALA151218-5G
4-(1,3-Benzothiazol-2-yl)phenylamine ≥97% (by GC, titration analysis)
4-(1,3-Benzothiazol-2-yl)phenylamine
Formula: C₁₃H₁₀N₂S
MW: 226.3 g/mol
Melting Pt: 151-158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00182896
CAS Number: 6278-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabca885d7fada86d1d5abf739bf29e26dccf168&vid=dabca885d7fada86d1d5abf739bf29e26dccf168&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR