2-Amino-4-(p-tolyl)thiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine, 4-(p-Tolyl)thiazol-2-amine, 2-Amino-4-(4-methylphenyl)thiazole

A151217-5G A151217-1G A151217-25G
ALA151217-5GEA 63.56 USD
ALA151217-5G ALA151217-1G ALA151217-25G
2-Amino-4-(p-tolyl)thiazole ≥98% (by HPLC)
2-Amino-4-(p-tolyl)thiazole
Formula: C₁₀H₁₀N₂S
MW: 190.26 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00170264
CAS Number: 2103-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38361781a52cfb090ad3c6a1600123457e916696&vid=38361781a52cfb090ad3c6a1600123457e916696&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR