2-Amino-4-methoxybenzothiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

A151216-1G A151216-5G
ALA151216-1GEA 40.76 USD
ALA151216-1G ALA151216-5G
2-Amino-4-methoxybenzothiazole ≥98% (by HPLC)
2-Amino-4-methoxybenzothiazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Melting Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00005792
CAS Number: 5464-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbdb8c37064621a15dd0bfd9cbced95982859096&vid=dbdb8c37064621a15dd0bfd9cbced95982859096&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR