2-Amino-4,5-dimethylthiazole hydrobromide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151214-1G A151214-25G A151214-5G
ALA151214-1GEA 9.41 USD
ALA151214-1G ALA151214-25G ALA151214-5G
2-Amino-4,5-dimethylthiazole hydrobromide ≥98%
2-Amino-4,5-dimethylthiazole hydrobromide
Formula: C₅H₉BrN₂S
Storage Temperature: Ambient
MDL Number: MFCD00035212
CAS Number: 7170-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b9085716bc9094ab50c2409ef2c27948dd6f352&vid=0b9085716bc9094ab50c2409ef2c27948dd6f352&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR