2-Amino-6-nitrobenzothiazole ≥96% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151211-1G A151211-5G
ALA151211-1GEA 12.26 USD
ALA151211-1G ALA151211-5G
2-Amino-6-nitrobenzothiazole ≥96% (by HPLC)
2-Amino-6-nitrobenzothiazole

Voltammetric behavior of 2-amino-6-nitrobenzothiazole has been investigated using direct current voltammetry and differential pulse voltammetry2-Amino-6-nitrobenzothiazole has been used: . as model analyte for voltammetric determination of electrochemically reducible organic substances[4] . in the synthesis of 2-methyl-4-nitro-2H-pyrazole-3-carboxylic acid[2-(cyclohexanecarbonylamino)benzothiazol-6-yl]amide derivatives . in the preparation of push-pull nonlinear optical chromophores containing thiazole and benzothiazole acceptors . as a base in dye production by diazotation reaction

Formula: C₇H₅N₃O₂S
Melting Pt: 247 - 249 °C
MDL Number: MFCD00005786
CAS Number: 6285-57-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=433a4276c49f3b92ec2505f309a613be67d80a2c&vid=433a4276c49f3b92ec2505f309a613be67d80a2c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR