5-Acetyl-2,4-dimethylthiazole ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2,4-Dimethyl-5-acetylthiazole, 1-(2,4-Dimethylthiazol-5-yl)ethanone, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-one

A151209-100G A151209-25G
ALA151209-100GEA 328.61 USD
ALA151209-100G ALA151209-25G
5-Acetyl-2,4-dimethylthiazole ≥98% (by GC)
5-Acetyl-2,4-dimethylthiazole
Formula: C₇H₉NOS
MW: 155.22 g/mol
Boiling Pt: 230°C
Density: 1.15 g/cm³
Flash Pt: 104°C
MDL Number: MFCD00005331
CAS Number: 38205-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b97c7875b5a9ffbf20d271cdd1055a4010814b0&vid=3b97c7875b5a9ffbf20d271cdd1055a4010814b0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR