2-(2-Aminothiazol-4-yl)acetic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazoleacetic acid

A151208-100G A151208-25G A151208-5G
ALA151208-100GEA 104.41 USD
ALA151208-100G ALA151208-25G ALA151208-5G
2-(2-Aminothiazol-4-yl)acetic acid ≥98% (by HPLC)
2-(2-Aminothiazol-4-yl)acetic acid
Formula: C₅H₆N₂O₂S
Melting Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010068
CAS Number: 29676-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd3219f3f91a4356f6fa199b06d200ece4755959&vid=bd3219f3f91a4356f6fa199b06d200ece4755959&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR