Acetone benzothiazolyl-2-hydrazone ≥97%
Supplier: ALADDIN SCIENTIFIC
A151207-1G
A151207-25G
A151207-5G
ALA151207-1GEA
16.06
USD
ALA151207-1G
ALA151207-25G
ALA151207-5G
Acetone benzothiazolyl-2-hydrazone ≥97%
Acetone benzothiazolyl-2-hydrazone
Formula:
C₁₀H₁₁N₃S Melting Pt: 198 °C |
MDL Number:
MFCD00059701 CAS Number: 6277-26-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...