2-Amino-6-methylbenzothiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151204-100G A151204-25G A151204-5G
ALA151204-100GEA 87.31 USD
ALA151204-100G ALA151204-25G ALA151204-5G
2-Amino-6-methylbenzothiazole ≥98% (by HPLC)
2-Amino-6-methylbenzothiazole

The anti-tetanus activity of 2-amino-6-methylbenzothiazole, a muscle relactant, was studied.2-Amino-6-methylbenzothiazole was used in the preparation of 2-[(6-methyl-1,3-benzothiazol-2-yl)amino]-N-[2-(substituted phenyl/furan-2-yl)-4-oxo-1,3-thiazolidin-3-yl]nicotinamides.

Formula: C₈H₈N₂S
MW: 164.23 g/mol
Melting Pt: 140-142°C
MDL Number: MFCD00005789
CAS Number: 2536-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f42f74bc7303bccbb88098a0954cddd17cd3b39b&vid=f42f74bc7303bccbb88098a0954cddd17cd3b39b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR