6-Amino-2-mercaptobenzothiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151203-100MG A151203-250MG
ALA151203100MGEA 11.31 USD
ALA151203100MG ALA151203250MG
6-Amino-2-mercaptobenzothiazole ≥97%
6-Amino-2-mercaptobenzothiazole
Formula: C₇H₆N₂S₂
Melting Pt: 267-272 °C
MDL Number: MFCD00059164
CAS Number: 7442-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR