2-Amino-4-methylthiazole hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151202-1G A151202-5G
ALA151202-1GEA 9.41 USD
ALA151202-1G ALA151202-5G
2-Amino-4-methylthiazole hydrochloride ≥95%
2-Amino-4-methylthiazole hydrochloride
Formula: C₄H₆N₂S
Melting Pt: 173 °C
MDL Number: MFCD00035214
CAS Number: 6142-15-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23412123bc93889e7c9adbddf6e031621be24d03&vid=23412123bc93889e7c9adbddf6e031621be24d03&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR