1-Allylhydantoin 98

Supplier: ALADDIN SCIENTIFIC
Warning

A151198-1G A151198-5G
ALA151198-1GEA 11.31 USD
ALA151198-1G ALA151198-5G
1-Allylhydantoin 98
1-Allylhydantoin
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00142580
CAS Number: 3366-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5674a4c4a1b86a396abf7573be1ca5e2528a9899&vid=5674a4c4a1b86a396abf7573be1ca5e2528a9899&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR