Allyl ether ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Diallyl Ether

A151191-100ML A151191-25ML A151191-5ML
ALA151191100MLEA 159.51 USD
ALA151191100ML ALA151191-25ML ALA151191-5ML
Allyl ether ≥98% (by GC)
Allyl ether

Allyl Ether is a modifier used in the manufacture of foamable polystyrene

Formula: C₆H₁₀O
Boiling Pt: 94°C
Density: 0.803 g/cm³
Flash Pt: -7°C
Storage Temperature: Refrigerator
MDL Number: MFCD00008650
CAS Number: 557-40-4
UN: 2360
ADR: 3, 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1639419125651db99b475127236de5dc370a4e39&vid=1639419125651db99b475127236de5dc370a4e39&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR