1-Adamantanecarboxamide ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151186-1G
A151186-5G
ALA151186-1GEA
40.76
USD
ALA151186-1G
ALA151186-5G
1-Adamantanecarboxamide ≥97% (by GC)
1-Adamantanecarboxamide
Formula:
C₁₁H₁₇NO Melting Pt: 186-190°C |
MDL Number:
MFCD00077200 CAS Number: 5511-18-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...