1-Adamantyl isocyanate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151185-200MG A151185-1G
ALA151185200MGEA 25.56 USD
ALA151185200MG ALA151185-1G
1-Adamantyl isocyanate ≥98%
1-Adamantyl isocyanate

1-Adamantyl isocyanate is an isocyanate derivative.

Formula: C₁₁H₁₅NO
Melting Pt: 144-149°C
Storage Temperature: Ambient
MDL Number: MFCD00134405
CAS Number: 4411-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6473ff23a547f47a82ac13c27c65720401b51e37&vid=6473ff23a547f47a82ac13c27c65720401b51e37&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR