1,3-Adamantanedicarboxylic acid ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1,3-Adamantanedicarboxylic Acid, 1,3-Dicarboxyadamantane, (1s,3s,5r,7r)-Adamantane-1,3-dicarboxylic acid

A151180-1G A151180-5G
ALA151180-1GEA 14.16 USD
ALA151180-1G ALA151180-5G
1,3-Adamantanedicarboxylic acid ≥97% (by GC)
1,3-Adamantanedicarboxylic acid
Formula: C₁₂H₁₆O₄
MW: 224.26 g/mol
MDL Number: MFCD00167790
CAS Number: 39269-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0d9b28facc7b29499a15435cb5841810374b9fc&vid=f0d9b28facc7b29499a15435cb5841810374b9fc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR