1,3-Adamantanediacetic acid ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1,3-Adamantanediacetic Acid, H2ADA, 1,3-Bis(carboxymethyl)adamantane, 1,3-Bis(carboxymethyl)adamantine

A151179-1G A151179-25G A151179-5G
ALA151179-1GEA 38.86 USD
ALA151179-1G ALA151179-25G ALA151179-5G
1,3-Adamantanediacetic acid ≥97% (by GC)
1,3-Adamantanediacetic acid
Formula: C₁₄H₂₀O₄
MW: 252.31 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00191708
CAS Number: 17768-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26ff7fbbc3acc6f0a64b217ac9ed16da99d17e4&vid=c26ff7fbbc3acc6f0a64b217ac9ed16da99d17e4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR