1-Acetamidoadamantane ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-1-Adamantylacetamide, N-Acetyl-1-adamantanamine

A151178-10G A151178-25G A151178-50G
ALA151178-10GEA 36.01 USD
ALA151178-10G ALA151178-25G ALA151178-50G
1-Acetamidoadamantane ≥98%
1-Acetamidoadamantane
Formula: C₁₂H₁₉NO
MW: 193.29 g/mol
Melting Pt: 148-149°C
Storage Temperature: Refrigerator
MDL Number: MFCD00074730
CAS Number: 880-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f2c716ceae6624771fd43ff65c0e9d1f91114ce&vid=0f2c716ceae6624771fd43ff65c0e9d1f91114ce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR