3-Amino-5,6-dimethyl-1,2,4-triazine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151175-100G A151175-1G A151175-25G A151175-5G
ALA151175-100GEA 188.96 USD
ALA151175-100G ALA151175-1G ALA151175-25G ALA151175-5G
3-Amino-5,6-dimethyl-1,2,4-triazine ≥98%
3-Amino-5,6-dimethyl-1,2,4-triazine

Product description 3-Amino-5,6-dimethyl-1,2,4-triazine reacts with silver trifluoro¬methane¬sulfonate to yield tris(3-amino-5,6-dimethyl-1,2,4-triazine-κN)silver(I) trifluromethane-sulfonate-3-amino-5,6-dimethyl-1,2,4-triazine. It reacts with an excess of benzoyl chloride in the presence of triethylamine in refluxing chloroform to give 3-dibenzoylamino-5,6-dimethyl-1,2,4-triazine.Application3-Amino-5,6-dimethyl-1,2,4-triazine was used in intramolecular cyclization reaction of various 1,2-bis(amidinohydrazone)s.

Formula: C₅H₈N₄
Melting Pt: 210 °C
MDL Number: MFCD00006460
CAS Number: 17584-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fc88b5b2978398c01283d8ca7949e20739e071e&vid=9fc88b5b2978398c01283d8ca7949e20739e071e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR