3-Aminopiperidine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Piperidin-3-amine

A151173-1G A151173-250MG A151173-5G
ALA151173-1GEA 150.96 USD
ALA151173-1G ALA151173250MG ALA151173-5G
3-Aminopiperidine ≥98% (by GC)
3-Aminopiperidine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
MDL Number: MFCD04972497
CAS Number: 54012-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfdd9107e98f95dcd90247b9cc565ecc5a8042d2&vid=cfdd9107e98f95dcd90247b9cc565ecc5a8042d2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR