2-Amino-6-(trifluoromethyl)benzothiazole ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
6-(Trifluoromethyl)benzo[d]thiazol-2-amine
A151170-1G
A151170-5G
ALA151170-1GEA
40.76
USD
ALA151170-1G
ALA151170-5G
2-Amino-6-(trifluoromethyl)benzothiazole ≥98% (by HPLC)
2-Amino-6-(trifluoromethyl)benzothiazole
Formula:
C₈H₅F₃N₂S MW: 218.20 g/mol Melting Pt: 115-124°C |
MDL Number:
MFCD00269597 CAS Number: 777-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...