SU 3327 ≥96%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole, SU 3327, 5-((5-Nitrothiazol-2-yl)thio)-1,3,4-thiadiazol-2-amine, SU3327

A151167-10MG A151167-1G A151167-250MG A151167-50MG
ALA151167-10MGEA 112.01 USD
ALA151167-10MG ALA151167-1G ALA151167250MG ALA151167-50MG
SU 3327 ≥96%, Moligand™
SU 3327

Product Introduction:SU3327 is a potent, selective and substrate-competitive JNK inhibitor with an IC50 of 0.7 μM. SU3327 also inhibits protein-protein interactions between JNK and JNK Interacting Protein (JIP) with an IC50 of 239 nM. SU3327 shows less active against p38α and Akt kinase.

Formula: C₅H₃N₅O₂S₃
MW: 261.30 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD01927019
CAS Number: 40045-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=200b6aa38bad32c662de0dc1ccdb85012a961b10&vid=200b6aa38bad32c662de0dc1ccdb85012a961b10&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR