α-Terpinene ≥90% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

A151162-100ML A151162-25ML
ALA151162100MLEA 88.26 USD
ALA151162100ML ALA151162-25ML
α-Terpinene ≥90% (by GC)
α-Terpinene

It is a volatile essential oil found in Melaleuca alternifolia. It has demonstrated antimicrobial properties against several human pathogens.A terpene with demonstrated antimicrobial properties.

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173 °C
Density: 0.84 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001534
CAS Number: 99-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc426b275db81208b2b845e9ea459a69b9c599a&vid=4bc426b275db81208b2b845e9ea459a69b9c599a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR