1-Amino-4-naphthol hydrochloride ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151160-5G
A151160-1G
ALA151160-5GEA
61.66
USD
ALA151160-5G
ALA151160-1G
1-Amino-4-naphthol hydrochloride ≥95%
1-Amino-4-naphthol hydrochloride
Formula:
C₁₀H₁₀ClNO Melting Pt: 280°C Storage Temperature: Ambient |
MDL Number:
MFCD00012562 CAS Number: 5959-56-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...