5-(Acetoacetamido)-2-benzimidazolinone ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 5-Acetoacetlamino benzimdazolone

A151157-100G A151157-25G
ALA151157-100GEA 159.51 USD
ALA151157-100G ALA151157-25G
5-(Acetoacetamido)-2-benzimidazolinone ≥98% (by HPLC)
5-(Acetoacetamido)-2-benzimidazolinone
Formula: C₁₁H₉N₃O₃
MW: 231.21 g/mol
MDL Number: MFCD00444508
CAS Number: 26576-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f5b685fa4a17381b3adcc93777e29174367d36b&vid=8f5b685fa4a17381b3adcc93777e29174367d36b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR