5-Amino-2-mercaptobenzimidazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Amino-1H-benzoimidazole-2-thiol

A151156-100G A151156-1G
ALA151156-100GEA 965.11 USD
ALA151156-100G ALA151156-1G
5-Amino-2-mercaptobenzimidazole ≥97%
5-Amino-2-mercaptobenzimidazole
Formula: C₇H₇N₃S
Melting Pt: 240-244°C
MDL Number: MFCD00022671
CAS Number: 2818-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea9b0d3f28a929673919ecfa811383a438c03a50&vid=ea9b0d3f28a929673919ecfa811383a438c03a50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR