α-Angelicalactone ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151155-100G A151155-25G A151155-500G A151155-5G
ALA151155-100GEA 144.31 USD
ALA151155-100G ALA151155-25G ALA151155-500G ALA151155-5G
α-Angelicalactone ≥95%
α-Angelicalactone

α-Angelica lactone, a cyclic lactone that occurs naturally in tobacco, is generally used in tobacco flavoring.

Formula: C₅H₆O₂
Boiling Pt: 56 °C
Melting Pt: 16 °C
Density: 1.1 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00005375
CAS Number: 591-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a2dbc8744fd0f3060ca51fb19b262ae949057ff&vid=3a2dbc8744fd0f3060ca51fb19b262ae949057ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR