5-Aminovaleric acid Hydroiodide, low water content ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151151-1G A151151-250MG
ALA151151-1GEA 114.86 USD
ALA151151-1G ALA151151250MG
5-Aminovaleric acid Hydroiodide, low water content ≥97%
5-Aminovaleric acid Hydroiodide
Formula: C₅H₁₁NO₂·HI
Storage Temperature: Ambient
CAS Number: 1705581-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e09697d4e3ef8c99b29c9625074d9bd5c07edf1&vid=0e09697d4e3ef8c99b29c9625074d9bd5c07edf1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR