Aniline Hydroiodide ≥98% (by HPLC, total nitrogen)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151150-1G
A151150-250MG
ALA151150-1GEA
66.41
USD
ALA151150-1G
ALA151150250MG
Aniline Hydroiodide ≥98% (by HPLC, total nitrogen)
Aniline Hydroiodide
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.
Formula:
C₆H₇N·HI Storage Temperature: Ambient |
CAS Number:
45497-73-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...