4-(4-Aminopiperidino)pyridine dihydrochloride ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride, 4-(4-Aminopiperidino)pyridine Dihydrochloride

A151147-1G A151147-200MG A151147-5G
ALA151147-1GEA 74.01 USD
ALA151147-1G ALA151147200MG ALA151147-5G
4-(4-Aminopiperidino)pyridine dihydrochloride ≥98% (by HPLC, titration analysis)
4-(4-Aminopiperidino)pyridine dihydrochloride
Formula: C₁₀H₁₅N₃
MW: 250.17 g/mol
MDL Number: MFCD06797043
CAS Number: 1169396-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6733aff7f68c810691fee52eeffdbff98b280dd&vid=d6733aff7f68c810691fee52eeffdbff98b280dd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR