4-Pentylpyridine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151146-1G A151146-250MG
ALA151146-1GEA 56.91 USD
ALA151146-1G ALA151146250MG
4-Pentylpyridine ≥95%
4-Pentylpyridine
Formula: C₁₀H₁₅N
Boiling Pt: 230 °C
Density: 0.91 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD02093422
CAS Number: 2961-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99852832e2cd6599feac102bf1bcbb1df30872fa&vid=99852832e2cd6599feac102bf1bcbb1df30872fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR